missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RCC1 Recombinant Rabbit Monoclonal Antibody (24GB4175)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen RCC1
Clone 24GB4175
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30274990
GREENER_CHOICE
View Documents
Invitrogen™
MA557215
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275445
View Documents
Invitrogen™
MA557216
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RCC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Guanine-nucleotide releasing factor that promotes the exchange of Ran-bound GDP by GTP. Involved in the regulation of onset of chromosome condensation in the S phase. Binds both to the nucleosomes and double-stranded DNA. RCC1-Ran complex (together with other proteins) acts as a component of a signal transmission pathway that detects unreplicated DNA. Plays a key role in nucleo-cytoplasmic transport, mitosis and nuclear-envelope assembly.
Specifications

Specifications

RCC1
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
RCC1
4931417M11Rik; AI326872; Cell cycle regulatory protein; chc1; chc1-a; chromosome condensation 1; chromosome condensation protein 1; fj77f08; guanine nucleotide-releasing protein; RanGEF; RCC1; rcc1.L; rcc1-a; rcc1-b; RCC1-I; Regulator of chromosome condensation; regulator of chromosome condensation 1; regulator of chromosome condensation 1 L homeolog; SNHG3-RCC1; SNHG3-RCC1 readthrough; wu:fj77f08; XELAEV_18012073mg; xrcc1; zgc:65796; zgc:77114
RCC1
Primary
Affinity chromatography
24GB4175
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P18754, Q8VE37
100088, 1104, 682908
A synthesized peptide derived from human RCC1 (aa 10-50).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.