missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCBTB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13225-1-AP-20UL
This item is not returnable.
View return policy
Description
RCBTB2 is a member of the RCC1-related GEF family. The N-terminal half of the amino acid sequence shows similarity to the regulator of chromosome condensation RCC1, which acts as a guanine nucleotide exchange factor (GEF) protein for the Ras-related GTPase Ran.Specifications
| RCBTB2 | |
| Polyclonal | |
| Unconjugated | |
| RCBTB2 | |
| CHC1 L, CHC1L, Chromosome condensation 1 like, RCBTB2, RCC1 like G exchanging factor, RLG | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 105670, 1102, 290363 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95199, Q6P798, Q99LJ7 | |
| Rcbtb2 | |
| RCBTB2 Fusion Protein Ag3975 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction