missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCBTB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522234
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (96%), Rat (96%), Xenopus laevis (83%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RCBTB2 is a member of the RCC1-related GEF family. The N-terminal half of the amino acid sequence shows similarity to the regulator of chromosome condensation RCC1, which acts as a guanine nucleotide exchange factor (GEF) protein for the Ras-related GTPase Ran.
Specifications
| RCBTB2 | |
| Polyclonal | |
| Unconjugated | |
| Rcbtb2 | |
| 2610028E02Rik; 2810420M18Rik; AI448345; AW240694; CHC1L; CHC1-L; Chromosome condensation 1-like; RC/BTB2; Rcbtb2; RCC1 and BTB domain containing protein 2; RCC1 and BTB domain-containing protein 2; RCC1-like G exchanging factor; RCC1-like G exchanging factor RLG; regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2; regulator of chromosome condensation and BTB domain-containing protein 2; RLG | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1102 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.32 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O95199 | |
| Rcbtb2 | |
| Recombinant protein encompassing a sequence within the center region of human RCBTB2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction