missing translation for 'onlineSavingsMsg'
Få mere at vide

RCBTB2 Antibody, Novus Biologicals™

Artikelnummer. 18332048 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.05 mg
Pakningsstørrelse:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
18332048 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18332048 Leverandør Novus Biologicals Leverandørnr. H00001102B01P

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Mouse Polyclonal Antibody

RCBTB2 Polyclonal antibody specifically detects RCBTB2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen RCBTB2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_001259.1
Gene Alias CHC1-L, CHC1LRCC1-like G exchanging factor RLG, Chromosome condensation 1-like, RCC1 and BTB domain-containing protein 2, RCC1-like G exchanging factor, regulator of chromosome condensation (RCC1) and BTB (POZ) domain containingprotein 2, regulator of chromosome condensation and BTB domain containing protein 2, Regulator of chromosome condensation and BTB domain-containing protein 2, RLG
Host Species Mouse
Immunogen RCBTB2 (NP_001259.1, 1 a.a. - 551 a.a.) full-length human protein. MEEELPLFSGDSGKPVQATLSSLKMLDVGKWPIFSLCSEEELQLIRQACVFGSAGNEVLYTTVNDEIFVLGTNCCGCLGLGDVQSTIEPRRLDSLNGKKIACLSYGSGPHIVLATTEGEVFTWGHNAYSQLGNGTTNHGLVPCHISTNLSNKQVIEVACGSYHSLVLTSDGEVFAWGYNNSGQVGSGSTVNQPIPRRVTGCLQNKVVVTIACGQMCCMAVVDTGEVYVWGYNGNGQLGLGNSGNQPTPCRVAALQGIRVQRVACGYAHTLVLTDEGQVYAWGANSYGQLGTGNKSNQSYPTPVTVEKDRIIEIAACHSTHTSAAKTQGGHVYMWGQCRGQSVILPHLTHFSCTDDVFACFATPAVTWRLLSVEPDDHLTVAESLKREFDNPDTADLKFLVDGKYIYAHKVLLKIRCEHFRSSLEDNEDDIVEMSEFSYPVYRAFLEYLYTDSISLSPEEAVGLLDLATFYRENRLKKLCQQTIKQGICEENAIALLSAAVKYDAQDLEEFCFRFCINHLTVVTQTSGFAEMDHDLLKNFISKASRVGAFKN
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1102
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.