missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCAS1 Monoclonal Antibody (K40051_4H7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556366
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: TLPTTVDYSS VPKQTDVEEW TSWDEDAPTS VKIEGGNGNV ATQQNSLEQL EPDYFKDMTP TIRKTQKIVI KKREPLNFGI PDGSTGFSSR LAATQDLPFI HQSSELGDLD TWQENTNAWE EEEDAAWQAE EVLRQQKLAD REKRAAEQQR KKMEKEAQRL MKKEQNKIGV
This gene was identified as an estrogen-responsive gene. Regulation of transcription by estrogen is mediated by estrogen receptor which binds to the estrogen-responsive element found in the 5'-flanking region of this gene. The encoded protein is a tumor-associated antigen that is expressed at high frequency in a variety of cancers. Two transcript variants differing in the 5' UTR, but encoding the same protein, have been identified for this gene.
Specifications
| RCAS1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O00559 | |
| EBAG9 | |
| Recombinant human EBAG9 (immunogen range is 41-210). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K40051_4H7 | |
| Unconjugated | |
| EBAG9 | |
| AI835379; BAG9; cancer associated surface antigen; cancer associated surface antigen RCAS1; cancer-associated surface antigen RCAS1; EB9; EBAG9; estrogen receptor binding fragment associated gene 9; estrogen receptor binding site associated, antigen, 9; estrogen receptor-binding fragment-associated gene 9; estrogen receptor-binding fragment-associated gene 9 protein; HGNC:3123; PDAF; Rcas1; receptor binding cancer antigen expressed on SiSo cells; Receptor-binding cancer antigen expressed on SiSo cells | |
| Mouse | |
| Protein A | |
| RUO | |
| 9166 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction