missing translation for 'onlineSavingsMsg'
Learn More

RCAN3 Antibody (OTI1B2), DyLight 680, Novus Biologicals™

Product Code. 18190147 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18190147 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18190147 Supplier Novus Biologicals Supplier No. NBP273842FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RCAN3 Monoclonal specifically detects RCAN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCAN3
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1B2
Conjugate DyLight 680
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias Down syndrome candidate region 1-like 2, Down syndrome critical region gene 1-like 2, DSCR1L2regulator of calcineurin 3 isoform 13,45,MCIP3regulator of calcineurin 3 isoform 1a2,34,5, hRCN3, Myocyte-enriched calcineurin-interacting protein 3, RCAN family member 3, RCN3, Regulator of calcineurin 3, regulator of calcineurin 3 isoform 1a3,45,Down syndrome candidate region 1-like protein 2, regulator of calcineurin 3 isoform 1b2,34,5, regulator of calcineurin 3 isoform 1c2,34,5, regulator of calcineurin 3 isoform 1c3,45,calcipressin-3
Gene Symbols RCAN3
Host Species Mouse
Immunogen Full length recombinant protein of human RCAN3 (NP_038469) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 11123.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.