missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCAN3 Antibody (OTI1B2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73842CL1
This item is not returnable.
View return policy
Description
RCAN3 Monoclonal antibody specifically detects RCAN3 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| RCAN3 | |
| Monoclonal | |
| CoraFluor 1 | |
| Down syndrome candidate region 1-like 2, Down syndrome critical region gene 1-like 2, DSCR1L2regulator of calcineurin 3 isoform 13,45,MCIP3regulator of calcineurin 3 isoform 1a2,34,5, hRCN3, Myocyte-enriched calcineurin-interacting protein 3, RCAN family member 3, RCN3, Regulator of calcineurin 3, regulator of calcineurin 3 isoform 1a3,45,Down syndrome candidate region 1-like protein 2, regulator of calcineurin 3 isoform 1b2,34,5, regulator of calcineurin 3 isoform 1c2,34,5, regulator of calcineurin 3 isoform 1c3,45,calcipressin-3 | |
| Full length human recombinant protein of human RCAN3(NP_038469) produced in E.coli. | |
| 0.1 mL | |
| Neuroscience | |
| 11123 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI1B2 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction