missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCAN3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-84972
This item is not returnable.
View return policy
Description
RCAN3 Polyclonal specifically detects RCAN3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RCAN3 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Down syndrome candidate region 1-like 2, Down syndrome critical region gene 1-like 2, DSCR1L2regulator of calcineurin 3 isoform 13,45,MCIP3regulator of calcineurin 3 isoform 1a2,34,5, hRCN3, Myocyte-enriched calcineurin-interacting protein 3, RCAN family member 3, RCN3, Regulator of calcineurin 3, regulator of calcineurin 3 isoform 1a3,45,Down syndrome candidate region 1-like protein 2, regulator of calcineurin 3 isoform 1b2,34,5, regulator of calcineurin 3 isoform 1c2,34,5, regulator of calcineurin 3 isoform 1c3,45,calcipressin-3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| RCAN3 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LRDTMKSWNDSQSDLCSTDQEEEEEMIFGENEDDLDEMMDLSDLPTSLFACSVHEAVFEAREQKERFEALFTIYDD | |
| 0.1 mL | |
| Neuroscience | |
| 11123 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction