missing translation for 'onlineSavingsMsg'
Learn More

RCAN2 Antibody, FITC, Novus Biologicals™

Product Code. 18410131 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410131 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410131 Supplier Novus Biologicals Supplier No. NBP176995F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RCAN2 Polyclonal antibody specifically detects RCAN2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCAN2
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias Calcipressin 2 (Thyroid hormone-responsive protein ZAKI-4) (Down syndromecandidate region 1-like 1) (Myocyte-enriched calcineurin interacting protein2) (MCIP2), calcipressin-2, Down syndrome candidate region 1-like 1, Down syndrome critical region gene 1-like 1, DSCR1L1MCIP2ZAKI4RCN2, hRCN2, Myocyte-enriched calcineurin-interacting protein 2, regulator of calcineurin 2CSP2, thyroid hormone-responsive (skin fibroblasts), Thyroid hormone-responsive protein ZAKI-4, ZAKI-4
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the center of human RCAN2. The immunogen is located within amino acids 70 - 120 of RCAN2. Amino Acid Squence: AQVQTPETDGDKLH
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10231
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.