missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCAN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586706
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
The RCAN1 protein interacts with calcineurin A and inhibits calcineurin-dependent signaling pathways, possibly affecting central nervous system development. This gene is located in the minimal candidate region for the Down syndrome phenotype, and is overexpressed in the brain of Down syndrome fetuses.
Specifications
| RCAN1 | |
| Polyclonal | |
| Unconjugated | |
| Rcan1 | |
| Adapt78; calcineurin inhibitor; calcipressin-1; calcium and oxidant-inducible mRNA; CALP1L; CSP1; Down syndrome candidate region 1; Down syndrome critical region gene 1; Down syndrome critical region homolog 1; Down syndrome critical region protein 1; Down syndrome critical region protein 1 homolog; DSC1; Dscr1; MCIP1; modulatory calcineurin-interacting protein 1; myocyte-enriched calcineurin-interacting protein 1; near DSCR proline-rich protein; Rcan1; RCN1; Regulator of calcineurin 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1827, 266766, 54720 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P53805, Q6IN33, Q9JHG6 | |
| Rcan1 | |
| Synthetic peptide, corresponding to the 207-252 of Human Calcipressin-1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction