missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCAN1 Monoclonal Antibody (OTI10C6)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527304
This item is not returnable.
View return policy
Description
RCAN1 Monoclonal Antibody for Western Blot
The RCAN1 protein interacts with calcineurin A and inhibits calcineurin-dependent signaling pathways, possibly affecting central nervous system development. This gene is located in the minimal candidate region for the Down syndrome phenotype, and is overexpressed in the brain of Down syndrome fetuses.
Specifications
| RCAN1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P53805, Q9JHG6 | |
| Rcan1 | |
| Full length human recombinant protein of RCAN1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI10C6 | |
| Unconjugated | |
| Rcan1 | |
| Adapt78; calcineurin inhibitor; calcipressin-1; calcium and oxidant-inducible mRNA; CALP1L; CSP1; Down syndrome candidate region 1; Down syndrome critical region gene 1; Down syndrome critical region homolog 1; Down syndrome critical region protein 1; Down syndrome critical region protein 1 homolog; DSC1; Dscr1; MCIP1; modulatory calcineurin-interacting protein 1; myocyte-enriched calcineurin-interacting protein 1; near DSCR proline-rich protein; Rcan1; RCN1; Regulator of calcineurin 1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1827, 54720 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction