missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCAN1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46853AF700
This item is not returnable.
View return policy
Description
RCAN1 Polyclonal specifically detects RCAN1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RCAN1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| P53805 | |
| RCAN1 | |
| Synthetic peptide made to an internal portion of human RCAN1 (within residues 210-252). [Swiss-Prot# P53805] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Immunogen has 92% identity to rat. | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ADAPT78, calcipressin-1, calcium and oxidant-inducible mRNA, Down syndrome candidate region 1, Down syndrome critical region gene 1, Down syndrome critical region protein 1, DSC1, modulatory calcineurin-interacting protein 1, Myocyte-enriched calcineurin-interacting protein 1, near DSCR proline-rich protein, regulator of calcineurin 1CSP1DSCR1MCIP1RCN1 | |
| Rabbit | |
| 38 kDa | |
| 0.1 mL | |
| Cancer, Neuroscience, Signal Transduction | |
| 1827 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction