missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RC74 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA566872

Product Code. 15949554

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: QPTSGKKRKRV SDDVPDCKVL KPLLSGSIPV EQFVQTLEKH GFSDIKVEDT AKGHIVLLQE AETLIQIEED STHIICDNDE MLRVRLRDLV LKFLQKF Highest antigen sequence identity to the following orthologs - mouse 94%, rat 95%.

INTS9 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A)and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1)and U2 (RNU2)(Baillat et al., 2005).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RC74
Polyclonal
Unconjugated
INTS9
BC028953; CPSF2L; D14Ertd231e; INT9; integrator complex subunit 9; INTS9; Protein related to CPSF subunits of 74 kDa; RC74; RC-74
Rabbit
Antigen affinity chromatography
RUO
55756
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NV88
INTS9
Recombinant Human RC74. Recombinant protein control fragment (Product #RP-105407).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.