missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RC74 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531305
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (99%), Zebrafish (84%), Xenopus laevis (87%), Chicken (92%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
INTS9 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A)and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1)and U2 (RNU2)(Baillat et al., 2005).
Specifications
| RC74 | |
| Polyclonal | |
| Unconjugated | |
| INTS9 | |
| BC028953; CPSF2L; D14Ertd231e; INT9; integrator complex subunit 9; INTS9; Protein related to CPSF subunits of 74 kDa; RC74; RC-74 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55756 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NV88 | |
| INTS9 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human RC74. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction