missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RC74 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93568-0.1ml
This item is not returnable.
View return policy
Description
RC74 Polyclonal antibody specifically detects RC74 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RC74 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| CPSF2L, FLJ10871, INT9, integrator complex subunit 9, Protein related to CPSF subunits of 74 kDa, RC74, RC-74 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 459-658 of human INTS9 (NP_060720.2). KLLKEVQPLHVVCPEQYTQPPPAQSHRMDLMIDCQPPAMSYRRAEVLALPFKRRYEKIEIMPELADSLVPMEIKPGISLATVSAVLHTKDNKHLLQPPPRPAQPTSGKKRKRVSDDVPDCKVLKPLLSGSIPVEQFVQTLEKHGFSDIKVEDTAKGHIVLLQEAETLIQIEEDSTHIICDNDEMLRVRLRDLVLKFLQKF | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55756 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction