missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBX1 Mouse anti-Human, Mouse, Rat, Clone: 1F4D9, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RBX1 |
|---|---|
| Clone | 1F4D9 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16848855
|
Proteintech
66716-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16838855
|
Proteintech
66716-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This locus encodes a RING finger-like domain-containing protein. The encoded protein interacts with cullin proteins and likely plays a role in ubiquitination processes necessary for cell cycle progression. This protein may also affect protein turnover. Related pseudogenes exist on chromosomes 2 and 5.Specifications
| RBX1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| BA554C12.1, Protein ZYP, RBX1, Regulator of cullins 1, ring box 1, RING box protein 1, RING finger protein 75, RNF75, ROC1 | |
| Rbx1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1F4D9 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Rat, Mouse | |
| P62877, P62878 | |
| 300084, 56438, 9978 | |
| RBX1 Fusion Protein Ag7005 | |
| Primary | |
| -20°C | |
| RBX1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title