missing translation for 'onlineSavingsMsg'
Learn More

RBX1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£292.00 - £435.00

Specifications

Antigen RBX1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18445071
View Documents
Novus Biologicals
NBP1-88742-25ul
25 μL
£292.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18272858
View Documents
Novus Biologicals
NBP1-88742
0.1 mL
£435.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RBX1 Polyclonal specifically detects RBX1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

RBX1
Polyclonal
Rabbit
Cancer, Cell Cycle and Replication, Hypoxia, Neuroscience, Wnt Signaling Pathway, Zinc Finger
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
9978
This antibody was developed against Recombinant Protein corresponding to amino acids:MAAAMDVDTPSGTNSGAGKKRFEVKKWNAVALWAWDIVVDNCAICRNHIMDLCIECQANQASATSEECTVAWGVCNHAFHFHCISRWLKTRQVCPLDNREWEF
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human
BA554C12.1, EC 6.3.2.-, FLJ60363, MGC13357, Protein ZYP, Rbx1, Regulator of cullins 1, RING box protein 1, RING finger protein 75, ring-box 1, ring-box 1, E3 ubiquitin protein ligase, RING-box protein 1, RNF75MGC1481, ROC1E3 ubiquitin-protein ligase RBX1, ZYP protein
RBX1
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Certificates

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.