missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBPSUHL Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-84239-0.1ML
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
RBPSUHL Polyclonal specifically detects RBPSUHL in Human samples. It is validated for Western Blot.
Especificaciones
| RBPSUHL | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 11317 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| RBPL, RBP-L, RBPSUHL, recombination signal binding protein for immunoglobulin kappa J region-like, recombining binding protein suppressor of hairless (Drosophila)-like, recombining binding protein suppressor of hairless-like protein, SUH, SUHL, Transcription factor RBP-L | |
| The immunogen is a synthetic peptide directed towards the N terminal region of human RBPSUHL | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido