missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBPMS Monoclonal Antibody (OTI3B7)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526396
This item is not returnable.
View return policy
Description
RBPMS Monoclonal Antibody for Western Blot, IHC (P), Flow
This gene encodes a member of the RRM family of RNA-binding proteins. The RRM domain is between 80-100 amino acids in length and family members contain one to four copies of the domain. The RRM domain consists of two short stretches of conserved sequence called RNP1 and RNP2, as well as a few highly conserved hydrophobic residues. The protein encoded by this gene has a single, putative RRM domain in its N-terminus. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| RBPMS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q93062 | |
| Rbpms | |
| Full length human recombinant protein of RBPMS produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3B7 | |
| Unconjugated | |
| Rbpms | |
| 2010300K22Rik; 2700019M19Rik; AU017537; Heart and RRM expressed sequence; HEart, RRM Expressed Sequence; HERMES; Rbpms; RBP-MS; RGD1561067; RNA binding protein gene with multiple splicing; RNA binding protein with multiple splicing; RNA binding protein, mRNA processing factor; RNA-binding protein gene with multiple splicing; RNA-binding protein with multiple splicing; RNA-binding protein with multiple splicing (RBP-MS) | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11030, 498642 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction