missing translation for 'onlineSavingsMsg'
Learn More

RBPMS Antibody (OTI3B7), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18426357 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426357 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426357 Supplier Novus Biologicals Supplier No. NBP273836MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RBPMS Monoclonal antibody specifically detects RBPMS in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBPMS
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3B7
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias FLJ32971, Heart and RRM expressed sequence, Hermes, HERMESRNA-binding protein with multiple splicing, RBP-MS, RNA binding protein with multiple splicing
Host Species Mouse
Immunogen Full length human recombinant protein of human RBPMS (NP_001008712) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Mismatch Repair
Primary or Secondary Primary
Gene ID (Entrez) 11030
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.