missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBPJL Polyclonal Antibody

RBPJL Polyclonal Antibody
Brand: Invitrogen PA598973
This item is not returnable.
View return policy
Description
RBPJL Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| RBPJL | |
| Polyclonal | |
| Unconjugated | |
| RBPJL | |
| RBP-J kappa-like; RBPJL; RBPL; RBP-L; RBPSUHL; recombination signal binding protein for immunoglobulin kappa J region like; recombination signal binding protein for immunoglobulin kappa J region-like; recombining binding protein suppressor of hairless-like protein; SUHL; Transcription factor RBP-L | |
| Rabbit | |
| Protein G | |
| RUO | |
| 11317 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.63 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UBG7 | |
| RBPJL | |
| Recombinant Human Recombining binding protein suppressor of hairless-like protein (1-151AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction