Learn More
Invitrogen™ RBPJ Monoclonal Antibody (6G4)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA549168
Product Code. 17941201
  • £362.00 / 100µg
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HEK293 whole cell, human HEPG2 whole cell, human HELA whole cell, human MCF-7 whole cell, human U87 whole cell, human HL-60 whole cell, rat NRK whole cell, mouse smooth muscle tissue. Flow: HEPA1-6 cell, U937 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
CSL plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair. The protein is abundantly expressed in heart and skeletal muscle and expression is increased by passive stretch. Mutations in this gene can lead to X-linked deafness.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RBPJ
Monoclonal
500 μg/mL
PBS with 4mg trehalose and no preservative
P31266, Q06330
RBPJ
E.coli-derived human RBPJK/RBPJ recombinant protein (Position: K41-Q467).
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2b
Flow Cytometry, Western Blot
6G4
Unconjugated
RBPJ
AI843960; AOS3; CBF1; CBF-1; csl; H-2K binding factor-2; Igkjrb; Igkjrb1; Igkrsbp; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; KBF2; Rbpj; RBP-J; RBP-J kappa; RBPJK; RBP-JK; rbp-jkappa; RBPSUH; recombination signal binding protein for immunoglobulin kappa J region; recombining binding protein suppressor of hairless; recombining binding protein suppressor of hairless-like; recombining binding protein suppressor of hairless-like protein; renal carcinoma antigen NY-REN-30; SUH; suppressor of hairless homolog; Unknown (protein for MGC:128782)
Mouse
Antigen affinity chromatography
RUO
19664, 3516, 679028
-20°C
Lyophilized