missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £451.00
Specifications
| Antigen | RBP7 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18416320
|
Novus Biologicals
NBP1-84383-25ul |
25 μL |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18755413
|
Novus Biologicals
NBP1-84383 |
0.1 mL |
£451.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RBP7 Polyclonal specifically detects RBP7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| RBP7 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| Cellular retinoic acid-binding protein 4, Cellular retinoic acid-binding protein IV, CRABP4, CRABP-IV, CRBP4, CRBPIV, MGC70641, putative cellular retinol-binding protein CRBP IV, retinoid binding protein 7, retinoid-binding protein 7, retinol binding protein 7, cellular | |
| RBP7 | |
| IgG | |
| Affinity Purified | |
| Specificity of human RBP7 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 116362 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RKIAKLLKPQKVIEQNGDSFTIHTNSSLRNYFVKFKVGEEFDEDNRGLDNRKCKSLVIWDNDRLTCIQKGEK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title