missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP5 Rabbit anti-Human, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11843-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| RBP5 | |
| Polyclonal | |
| Unconjugated | |
| RBP5 | |
| CRBP III, CRBP3, CRBPIII, HRBPiso, RBP5, Retinol binding protein 5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100270724, 83758 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P82980 | |
| RBP5 | |
| RBP5 Fusion Protein Ag2485 | |
| 20 μL | |
| Primary | |
| Pig, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction