missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBP5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145331
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a cellular retinol-binding protein expressed highly in kidney and liver. Down-regulation of the encoded protein in hepatocellular carcinoma was associated with large tumor size and poor patient survival rates.
Specifications
| RBP5 | |
| Polyclonal | |
| Unconjugated | |
| RBP5 | |
| cellular retinol binding protein 5; cellular retinol-binding protein III; CRBP3; CRBPIII; CRBP-III; HRBPiso; putative cellular retinol-binding protein CRBP III; RBP5; retinol binding protein 5; retinol binding protein 5, cellular; retinol-binding protein 5; retinol-binding protein 5, cellular | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100270724, 539418 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P82708 | |
| RBP5 | |
| Recombinant Bovine RBP5 protein, Met1-Lys135 (Accession #P82708). | |
| 200 μL | |
| Primary | |
| Bovine, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction