missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106491
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RBP5.
The protein encoded by this gene is a cellular retinol-binding protein expressed highly in kidney and liver. Down-regulation of the encoded protein in hepatocellular carcinoma was associated with large tumor size and poor patient survival rates.
Specifications
| RBP5 | |
| Polyclonal | |
| Unconjugated | |
| RBP5 | |
| cellular retinol binding protein 5; cellular retinol-binding protein III; CRBP3; CRBPIII; CRBP-III; HRBPiso; putative cellular retinol-binding protein CRBP III; RBP5; retinol binding protein 5; retinol binding protein 5, cellular; retinol-binding protein 5; retinol-binding protein 5, cellular | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83758 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P82980 | |
| RBP5 | |
| A synthesized peptide derived from human RBP5(Accession P82980), corresponding to amino acid residues G80-V130. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction