missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBP4 Recombinant Rabbit Monoclonal Antibody (9G7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560664
This item is not returnable.
View return policy
Description
RBP4 Recombinant Monoclonal Antibody for IHC (P), ELISA
This protein belongs to the lipocalin family and is the specific carrier for retinol in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein post translationally and results in defective delivery and supply to the epidermal cells.
Specifications
| RBP4 | |
| Recombinant Monoclonal | |
| 0.225 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P02753 | |
| RBP4 | |
| A synthesized peptide derived from human RBP4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 9G7 | |
| Unconjugated | |
| RBP4 | |
| MCOPCB10; Plasma retinol binding protein 4 (PRBP); Plasma retinol-binding protein; Plasma retinol-binding protein(1-176); Plasma retinol-binding protein(1-179); Plasma retinol-binding protein(1-181); Plasma retinol-binding protein(1-182); PRBP; PRO2222; RBP; Rbp4; Rbp-4; RBPA; RDCCAS; retinol; retinol binding; retinol binding protein 4; Retinol binding protein 4 interstitial; retinol binding protein 4, cellular; retinol binding protein 4, plasma; retinol-binding protein 4; retinol-binding protein 4, interstitial | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5950 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction