missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP4/Retinol-Binding Protein 4 Antibody (4C2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37614-0.025ml
This item is not returnable.
View return policy
Description
RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RBP4/Retinol-Binding Protein 4 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| RBP4 | |
| Purified recombinant fragment of human RBP expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 4°C2 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma | |
| Mouse | |
| 23 kDa | |
| 0.025 mL | |
| Cancer, Diabetes Research, Lipid and Metabolism | |
| 5950 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction