missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sino Biological™ RBP4 Recombinant Human Protein
Expressed from DNA sequence encoding human RBP4, including signal sequence (Met 1-Leu 201), fused to polyhistidine tag at C-terminus
Brand: Sino Biological™ 10354H08H25
This item is not returnable.
View return policy
Description
Expressed from DNA sequence encoding human RBP4, including signal sequence (Met 1-Leu 201), fused to polyhistidine tag at C-terminus
- Measured by its ability to bind all-trans retinoic acid
- Binding of retinoic acid results in quenching of tryptophan fluorescence in RBP4
- Recombinant human RBP4 can bind >1.0μM all-trans retinoic acid
- Lyophilized in PB
Specifications
5950 | |
5 x 50μg | |
Recombinant | |
Retinol binding protein 4, plasma | |
RBP4 Recombinant Human Protein |
23kd | |
Human | |
<1 EU/μg | |
RBP7 |
For Research Use Only. Not for use in diagnostic procedures.