missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93946-0.1ml
This item is not returnable.
View return policy
Description
RBP3 Polyclonal antibody specifically detects RBP3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RBP3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| D10S64, D10S65, D10S66, Interphotoreceptor retinoid-binding protein, Interstitial retinol-binding protein, IRBP, RBPI, retinol binding protein 3, interstitial, retinol-binding protein 3, retinol-binding protein 3, interstitial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 18-320 of human RBP3 (NP_002891.1). GPTHLFQPSLVLDMAKVLLDNYCFPENLLGMQEAIQQAIKSHEILSISDPQTLASVLTAGVQSSLNDPRLVISYEPSTPEPPPQVPALTSLSEEELLAWLQRGLRHEVLEGNVGYLRVDSVPGQEVLSMMGEFLVAHVWGNLMGTSALVLDLRHCTGGQVSGIPYIISYLHPGNTILHVDTIYNRPSNTTTEIWTLPQVLGERYGADKDVVVLTSSQTRGVAEDIAHILKQMRRAIVVGERTGGGALDLRKLRIGESDFFFTVPVSRSLGPLGGGSQTWEGSGVLPCVGTPAEQALEKALAIL | |
| 0.1 mL | |
| Neuroscience | |
| 5949 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction