Learn More
RBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22683-1-AP-20UL
 View more versions of this product
Product Code. 16852045
  • £135.00 / 20µL
Description
This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
RBP1
Polyclonal
Unconjugated
RBP1
CRABP I, CRBP, CRBP I, CRBP1, CRBPI, RBP1, RBPC, Retinol binding protein 1
Rabbit
Antigen Affinity Chromatography
RUO
19659, 25056, 5947
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02696, P09455, Q00915
Rbp1
RBP1 Fusion Protein Ag18572
20 μL
Primary
Rat, Mouse, Human