missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22683-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| RBP1 | |
| Polyclonal | |
| Unconjugated | |
| RBP1 | |
| CRABP I, CRBP, CRBP I, CRBP1, CRBPI, RBP1, RBPC, Retinol binding protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19659, 25056, 5947 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02696, P09455, Q00915 | |
| Rbp1 | |
| RBP1 Fusion Protein Ag18572 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction