missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP1 Monoclonal Antibody (G4E4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA135782
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-35782 only detects methylated RBP. In order to detect RBP, a methylation step of the antigen must be applied.
This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
RBP1 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.02% sodium azide | |
P02696, P09455, Q00915 | |
Rbp1 | |
Human plasma RBP. | |
100 μg | |
Primary | |
Goat, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
G4E4 | |
Unconjugated | |
Rbp1 | |
C CRABP-I; cellular retinol binding protein 1; Cellular retinol-binding protein; Cellular retinol-binding protein I; CRABP-I; CRBP; CRBP1; Crbpi; CRBP-I; LOW QUALITY PROTEIN: retinol-binding protein 1; mCRBPI; RBP; RBP1; Rbp-1; RBPC; retinol; retinol binding; Retinol Binding Protein; retinol binding protein 1; retinol binding protein 1, cellular; retinol-binding protein 1; retinol-binding protein 1, cellular | |
Mouse | |
purified | |
RUO | |
102189996, 19659, 25056, 5947 | |
4° C | |
Liquid |