missing translation for 'onlineSavingsMsg'
Learn More

RBMY1A1 Antibody (3G1), Novus Biologicals™

Product Code. 18342129 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342129 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342129 Supplier Novus Biologicals Supplier No. H00005940M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RBMY1A1 Monoclonal antibody specifically detects RBMY1A1 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBMY1A1
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3G1
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Alias MGC181956, RBM, RBM1, RBM2, RBMY, RBMY1C, RNA binding motif protein, Y chromosome, family 1, member A1, RNA binding motif protein, Y-linked, family 1, member A1, RNA binding protein, RNA-binding motif protein 1, RNA-binding motif protein 2, RNA-binding motif protein, Y chromosome, family 1 member A1/C, y chromosome RNA recognition motif 1, YRRM1, YRRM2
Host Species Mouse
Immunogen RBMY1A1 (NP_001007527.1, 257 a.a. ~ 356 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. YGTSHGAPPARGPRMSYGGSTCHAYSNTRDRYGRSWESYSSCGDFHYCDREHVCRKDQRNPPSLGRVLPDPREACGSSSYVASIVDGGESRSEKGDSSRY
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5940
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.