missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RBMXL2 Recombinant Rabbit Monoclonal Antibody (K01_1S37)

Product Code. 30828049
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828049 50 μL
30830127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828049 Supplier Invitrogen™ Supplier No. MA565302
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

RBMXL2 Recombinant Monoclonal Antibody for Western Blot

The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. RBMXL2 has two RRM domains that bind RNAs. RBMXL2 has similarity to HNRPG and RBMY proteins and it is suggested to replace HNRPG protein function during meiotic prophase or act as a germ cell-specific splicing regulator. It primarily localizes to the nuclei of meiotic spermatocytes. This gene is a candidate for autosomal male infertility.This gene belongs to the HNRPG subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBMXL2
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1S37
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene RBMXL2
Gene Accession No. O75526
Gene Alias 1700012H05Rik; Heterogeneous nuclear ribonucleoprotein G; heterogeneous nuclear ribonucleoprotein G T; hnRNP G; hnRNP G-T; HNRNPGT; Hnrnpg-t; HNRPG; HNRPGT; LOC617969 protein; NEWGENE_1598286; RBMX; RBMX like 2; Rbmxl2; RNA binding motif protein, X-linked like 2; RNA binding motif protein, X-linked-like 2; RNA-binding motif protein, X chromosome; RNA-binding motif protein, X chromosome, N-terminally processed; RNA-binding motif protein, X-linked-like-2; testes specific heterogenous nuclear ribonucleoprotein G T; testes-specific heterogenous nuclear ribonucleoprotein G-T; testis-specific heterogeneous nuclear ribonucleoprotein G-T
Gene Symbols RBMXL2
Host Species Rabbit
Immunogen Recombinant protein of human hnRNP G-T.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103690209, 27288, 76572
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.