missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBMX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599433
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total hnRNP G.
hnRNP G is a glycoprotein, which was originally identified as an autoantigen from German shepherd dogs with lupus-like syndrome. The gene encoding hnRNP G is located on chromosome Xq26 and is ubiquitously expressed. It contains one RNP-consensus RNA binding domain (RBD) and is related to RBMY, which is involved in spermatogenesis, and hnRNP G-T, which is a testis specific protein. All three proteins interact with Tra2b and therefore, are involved in pre-mRNA splicing.
Specifications
| RBMX | |
| Polyclonal | |
| Unconjugated | |
| RBMX | |
| glycoprotein p43; Heterogeneous nuclear ribonucleoprotein G; hnRNP G; HNRNPG; hnRNP-G; Hnrpg; MRXS11; RBMX; Rbmxp1; RBMXRT; RNA binding motif protein, X chromosome; RNA binding motif protein, X-linked; RNA-binding motif protein, X chromosome; RNA-binding motif protein, X chromosome retrogene; RNA-binding motif protein, X chromosome retrogene-like; RNA-binding motif protein, X chromosome, N-terminally processed; RNMX | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19655, 27316, 302855 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P38159, Q4V898, Q9WV02 | |
| RBMX | |
| A synthesized peptide derived from human RBMX(Accession P38159), corresponding to amino acid residues N20-N70. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction