missing translation for 'onlineSavingsMsg'
Learn More

RBMS2 Antibody (PCRP-RBMS2-1B6), DyLight 350, Novus Biologicals™

Product Code. 18339632 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339632 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339632 Supplier Novus Biologicals Supplier No. NBP314163UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RBMS2 Monoclonal antibody specifically detects RBMS2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBMS2
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-RBMS2-1B6
Conjugate DyLight 350
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ39093, FLJ40023, RNA binding motif, single stranded interacting protein 2, RNA-binding motif, single-stranded-interacting protein 2, SCR3FLJ43262, Suppressor of CDC2 with RNA-binding motif 3
Host Species Mouse
Immunogen Recombinant full-length human RBMS2 protein (Uniprot: Q15434)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5939
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.