missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBMS2 Antibody (PCRP-RBMS2-1B6), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14163AF750
This item is not returnable.
View return policy
Description
RBMS2 Monoclonal antibody specifically detects RBMS2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
| RBMS2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| FLJ39093, FLJ40023, RNA binding motif, single stranded interacting protein 2, RNA-binding motif, single-stranded-interacting protein 2, SCR3FLJ43262, Suppressor of CDC2 with RNA-binding motif 3 | |
| Recombinant full-length human RBMS2 protein (Uniprot: Q15434) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunofluorescence | |
| PCRP-RBMS2-1B6 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5939 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction