Learn More
RBMS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11061-2-AP-20UL
 View more versions of this product
Product Code. 16847883
  • £135.00 / 20µL
Description
This gene encodes a member of a small family of proteins which bind single stranded DNA/RNA. These proteins are characterized by the presence of two sets of ribonucleoprotein consensus sequence (RNP-CS) that contain conserved motifs, RNP1 and RNP2, originally described in RNA binding proteins, and required for DNA binding. These proteins have been implicated in such diverse functions as DNA replication, gene transcription, cell cycle progression and apoptosis. Several transcript variants, resulting from alternative splicing and encoding different isoforms, have been described. A pseudogene for this locus is found on chromosome 12.
Specifications
Specifications
RBMS1
Polyclonal
Unconjugated
RBMS1
C2orf12, HCC 4, MSSP, MSSP 1, MSSP 2, MSSP 3, MSSP1, RBMS1, SCR2, YC1
Rabbit
Antigen Affinity Chromatography
RUO
362138, 56878, 5937
-20°C
Liquid
Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P29558, Q5PQP1, Q91W59
Rbms1
RBMS1 Fusion Protein Ag1493
20 μL
Primary
Rat, Mouse, Human