missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBMS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20110
This item is not returnable.
View return policy
Description
RBMS1 Polyclonal specifically detects RBMS1 in Human samples. It is validated for Western Blot.
Specifications
| RBMS1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| chromosome 2 open reading frame 12, DKFZp564H0764, HCC-4, MGC15146, MGC3331, MGC97258, MGC97270, MGC97282, MGC99543, MSSP1, MSSP-1, MSSP-2, MSSP-3, RNA binding motif, single stranded interacting protein 1, SCR2MGC70597, single-stranded-interacting protein 1, suppressor of cdc 2 (cdc13) with RNA binding motif 2 | |
| Rabbit | |
| 45 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Bovine (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| P29558 | |
| RBMS1 | |
| Recombinant protein encompassing a sequence within the center region of human RBMS1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 5937 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction