Learn More
Invitrogen™ RBM8A Recombinant Rabbit Monoclonal Antibody (24GB1020)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557234
 View more versions of this product
Product Code. 30275318
  • £126.00 / 20µL
Description
RBM8A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a protein with a conserved RNA-binding motif. The protein is found predominantly in the nucleus, although it is also present in the cytoplasm. It is preferentially associated with mRNAs produced by splicing, including both nuclear mRNAs and newly exported cytoplasmic mRNAs. It is thought that the protein remains associated with spliced mRNAs as a tag to indicate where introns had been present, thus coupling pre- and post-mRNA splicing events. Previously, it was thought that two genes encode this protein, RBM8A and RBM8B; it is now thought that the RBM8B locus is a pseudogene. Two alternative start codons result in two forms of the protein, and this gene also uses multiple polyadenylation sites.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RBM8A
Recombinant Monoclonal
Unconjugated
RBM8A
2310057C03Rik; AA673428; Binder of OVCA1-1; BOV1; BOV-1; BOV1A; BOV-1A; BOV1B; BOV-1B; BOV1C; BOV-1C; C1DELq21.1; DEL1q21.1; fb34f03; HSPC114; MDS014; Rbm8; RBM8A; rbm8-A; rbm8a.S; rbm8a-A; rbm8a-b; RBM8B; ribonucleoprotein RBM8; ribonucleoprotein RBM8A; Ribonucleoprotein RBM8A-A; RNA binding motif protein 8; RNA binding motif protein 8A; RNA binding motif protein 8A S homeolog; RNA binding motif protein 8B; rna-binding; RNA-binding motif protein 8A; RNA-binding motif protein 8A-A; RNA-binding protein 8A; RNA-binding protein 8A-A; RNA-binding protein Y14; TAR; tsunagi; Unknown (protein for MGC:127566); wu:fb34f03; XELAEV_18042903mg; Y14; zgc:110525; ZNRP; zrnp1
Rabbit
Affinity chromatography
RUO
295284, 60365, 9939
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1020
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q27W01, Q9CWZ3, Q9Y5S9
RBM8A
A synthesized peptide derived from human RBM8A (aa 1-36).
20 μL
Primary
Human, Mouse, Rat