missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBM8A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585992
This item is not returnable.
View return policy
Description
RBM8A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a protein with a conserved RNA-binding motif. The protein is found predominantly in the nucleus, although it is also present in the cytoplasm. It is preferentially associated with mRNAs produced by splicing, including both nuclear mRNAs and newly exported cytoplasmic mRNAs. It is thought that the protein remains associated with spliced mRNAs as a tag to indicate where introns had been present, thus coupling pre- and post-mRNA splicing events. Previously, it was thought that two genes encode this protein, RBM8A and RBM8B; it is now thought that the RBM8B locus is a pseudogene. Two alternative start codons result in two forms of the protein, and this gene also uses multiple polyadenylation sites.
Specifications
| RBM8A | |
| Polyclonal | |
| Unconjugated | |
| RBM8A | |
| 2310057C03Rik; AA673428; Binder of OVCA1-1; BOV1; BOV-1; BOV1A; BOV-1A; BOV1B; BOV-1B; BOV1C; BOV-1C; C1DELq21.1; DEL1q21.1; fb34f03; HSPC114; MDS014; Rbm8; RBM8A; rbm8-A; rbm8a.S; rbm8a-A; rbm8a-b; RBM8B; ribonucleoprotein RBM8; ribonucleoprotein RBM8A; Ribonucleoprotein RBM8A-A; RNA binding motif protein 8; RNA binding motif protein 8A; RNA binding motif protein 8A S homeolog; RNA binding motif protein 8B; rna-binding; RNA-binding motif protein 8A; RNA-binding motif protein 8A-A; RNA-binding protein 8A; RNA-binding protein 8A-A; RNA-binding protein Y14; TAR; tsunagi; Unknown (protein for MGC:127566); wu:fb34f03; XELAEV_18042903mg; Y14; zgc:110525; ZNRP; zrnp1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 295284, 60365, 9939 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q27W01, Q9CWZ3, Q9Y5S9 | |
| RBM8A | |
| Synthetic peptide within Human Y14 aa 125-174. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction