missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBM6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555530
This item is not returnable.
View return policy
Description
Immunogen sequence: DFRDRDTPHS DFRGRHRSRT DQDFRGREMG SCMEFKDREM PPVDPNILDY IQPSTQDREH SGMNVNRREE STHDHTIERP AFGIQKGEFE H Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.
DEF-3 and LUCA15 belong to an evolutionarily conserved family of RNA binding proteins and share similar expression patterns. Both DEF-3 and LUCA15 are highly expressed in adult heart and thymus as well as fetal kidney. Conversely, fetal thymus and adult kidney express very little DEF-3 and LUCA15. In the haemopoietic system of mice, the expression of DEF-3 is downregulated upon differentiation of progenitor cells into granulocytes but persists during macrophage development. Both DEF-3 and LUCA15 contain two zinc finger motifs, a bipartite nuclear signal and two RNA binding motifs. DEF-3 and LUCA15 are capable of specifically binding poly(G) RNA.
Specifications
| RBM6 | |
| Polyclonal | |
| Unconjugated | |
| RBM6 | |
| 3G2; 4930506F14Rik; DEF3; DEF-3; ETK2; g16; HEK2; HLC-11; Lung cancer antigen NY-LU-12; lung cancer protooncogene 11; mKIAA4015; NY-LU-12; Protein G16; Rbm6; RGD1560367; RNA binding motif protein 6; RNA-binding motif protein 6; RNA-binding protein 6; RNA-binding protein DEF-3; TYRO6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10180 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P78332 | |
| RBM6 | |
| Recombinant protein corresponding to Human RBM6. Recombinant protein control fragment (Product #RP-95087). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction