missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM6 Antibody (4B3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00010180-M16
This item is not returnable.
View return policy
Description
RBM6 Monoclonal antibody specifically detects RBM6 in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| RBM6 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| DEF-3, DEF3FLJ39446,3G2, DKFZp686B0877, FLJ36517, FLJ42036, g16, HLC-11, Lung cancer antigen NY-LU-12, lung cancer protooncogene 11, NY-LU-12, Protein G16, RNA binding motif protein 6, RNA-binding motif protein 6, RNA-binding protein 6, RNA-binding protein DEF-3 | |
| RBM6 (NP_005768.1, 1024 a.a. ~ 1123 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DSPERKRIKYSRETDSDRKLVDKEDIDTSSKGGCVQQATGWRKGTGLGYGHPGLASSEEAEGRMRGPSVGASGRTSKRQSNETYRDAVRRVMFARYKELD | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA | |
| 4B3 | |
| Western Blot 1:500 | |
| NP_005768 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 10180 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction