missing translation for 'onlineSavingsMsg'
Learn More

RBM5 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18404228 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404228 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404228 Supplier Novus Biologicals Supplier No. NBP126613V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RBM5 Polyclonal antibody specifically detects RBM5 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM5
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias G15, H37, LUCA15FLJ39876, Protein G15, Putative tumor suppressor LUCA15, Renal carcinoma antigen NY-REN-9, RMB5, RNA binding motif protein 5, RNA-binding motif protein 5, RNA-binding protein 5
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 765 and 815 of human RNA binding motif protein 5 using the numbering given in entry NP_005769.1 (GeneID 10181).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10181
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.