missing translation for 'onlineSavingsMsg'
Learn More

RBM4B Antibody, Novus Biologicals™

Product Code. p-7110026 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18277195 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18277195 Supplier Novus Biologicals Supplier No. NBP180469

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RBM4B Polyclonal specifically detects RBM4B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM4B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_113680
Gene Alias MGC10871, RBM30, RBM4L, RNA binding motif protein 30, RNA binding motif protein 4B, RNA-binding motif protein 30, RNA-binding motif protein 4B, RNA-binding protein 30, RNA-binding protein 4B, ZCCHC15, ZCRB3B, zinc finger CCHC-type and RNA binding motif 3B
Gene Symbols RBM4B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal of human RBM4B. Peptide sequence AAAATTSSYYGRDRSPLRRAAAMLPTVGEGYGYGPESELSQASAATRNSL.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83759
Test Specificity Expected identity based on immunogen sequence: Bovine: 100%; Human: 100%; Pig: 100%; Canine: 92%; Mouse: 85%; Rat: 85%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.