missing translation for 'onlineSavingsMsg'
Learn More

RBM4B Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-93750-0.1ml

 View more versions of this product

Product Code. 18645290

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

RBM4B Polyclonal antibody specifically detects RBM4B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RBM4B
Polyclonal
Western Blot 1:500-1:2000
MGC10871, RBM30, RBM4L, RNA binding motif protein 30, RNA binding motif protein 4B, RNA-binding motif protein 30, RNA-binding motif protein 4B, RNA-binding protein 30, RNA-binding protein 4B, ZCCHC15, ZCRB3B, zinc finger CCHC-type and RNA binding motif 3B
Recombinant fusion protein containing a sequence corresponding to amino acids 140-359 of human RBM4B (NP_113680.1). RMHVQLSTSRLRTAPGMGDQSGCYRCGKEGHWSKECPVDRTGRVADFTEQYNEQYGAVRTPYTMGYGESMYYNDAYGALDYYKRYRVRSYEAVAAAAAASAYNYAEQTMSHLPQVQSTTVTSHLNSTSVDPYDRHLLPNSGAAATSAAMAAAAATTSSYYGRDRSPLRRAAAMLPTVGEGYGYGPESELSQASAATRNSLYDMARYEREQYVDRARYSAF
0.1 mL
Primary
Human
Purified
Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
83759
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.