missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM46 Antibody (OTI1D6), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73833DL594
This item is not returnable.
View return policy
Description
RBM46 Monoclonal antibody specifically detects RBM46 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| RBM46 | |
| Monoclonal | |
| DyLight 594 | |
| Cancer/testis antigen 68, CT68MGC27016, probable RNA-binding protein 46, RNA binding motif protein 46, RNA-binding motif protein 46 | |
| Full length recombinant protein of human RBM46 (NP_659416) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| OTI1D6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 166863 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction