missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM43 Antibody (S03-6I5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19816-100ul
This item is not returnable.
View return policy
Description
RBM43 Monoclonal antibody specifically detects RBM43 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Sandwich ELISA
Specifications
| RBM43 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 375287 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| S03-6I5 | |
| Unconjugated | |
| C2orf38, FLJ45645, RNA binding motif protein 43, RNA-binding motif protein 43, RNA-binding protein 43 | |
| Recombinant protein of human RBM43 (Uniprot # Q6ZSC3) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction