missing translation for 'onlineSavingsMsg'
Learn More

RBM42 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£143.00 - £251.00

Specifications

Antigen RBM42
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18610629
View Documents
Novus Biologicals
NBP3-12649-100ul
100 μg
£251.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18610839
View Documents
Novus Biologicals
NBP3-12649-50ul
50 μg
£143.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RBM42 Polyclonal antibody specifically detects RBM42 in Human samples. It is validated for Western Blot
Specifications

Specifications

RBM42
Western Blot
Unconjugated
Rabbit
Human
MGC10433, RNA binding motif protein 42, RNA-binding motif protein 42, RNA-binding protein 42
Produced in rabbits immunized with E. coli-derived Human RBM42 fragment
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
PBS, pH7.0
79171
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.